Learn More
Bio X Cell Antibody, Anti-Mouse PD-1 (CD279), 29F.1A12, 50 MG
Supplier: Bio X Cell BP0273R050MG
The 29F.1A12™ monoclonal antibody reacts with mouse PD-1 (programmed death-1), also known as CD279. PD-1 is a cell surface receptor that negatively regulates TCR signals by binding to its ligands, PD-L1 and PD-L2, inhibiting T-cell activation, proliferation, and cytokine production.
- Isotype: Rat IgG2a
- Conjugation: Unconjugated
- Immunogen: Recombinant PD-1-Ig fusion protein
- Formulation: PBS, pH 7.0, no stabilizers or preservatives
- Endotoxin: ≤0.5 EU/mg
- Aggregation: <5%
- Purity: ≥95%
- Sterility: 0.2 µm filtration
- Production: Purified from animal-free cell culture
- Purification: Protein G
- Molecular weight: 150 kDa
- Applications: In vivo blocking of PD-1/PD-L signaling, in vitro PD-1 neutralization, IHC (frozen), IF, Western blot, Flow cytometry, in vitro organoids
- Storage: Store at 4°C; do not freeze
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.