Learn More
Bio X Cell InVivoMAb anti-mouse PD-1 (CD279) Antibody, 5 MG
Supplier: Bio X Cell BE0273R005MG
The InVivoMAb anti-mouse PD-1 (CD279) antibody is designed for in vivo blocking of PD-1/PD-L signaling and in vitro PD-1 neutralization, making it suitable for various research applications.
- Applications include in vivo blocking of PD-1/PD-L signaling, in vitro PD-1 neutralization, immunohistochemistry (frozen), immunofluorescence, western blot, and flow cytometry
- Host species: rat
- Species reactivity: mouse
- Isotype: IgG2a
- Immunogen: recombinant PD-1-Ig fusion protein
- Store at 4°C; do not freeze
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.