Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bio X Cell Anti-Mouse PD-1 (CD279), Clone J43, 5 MG

Supplier:  Bio X Cell BE00332R005MG

Encompass_Preferred

The J43 monoclonal antibody targets mouse PD-1 (CD279), a 50-55 kDa cell surface receptor crucial for regulating T-cell activation. It belongs to the CD28 family of the immunoglobulin superfamily and is expressed on various immune cells. This antibody has demonstrated efficacy in blocking the binding of both mouse PD-L1-Ig and mouse PD-L2-Ig to PD-1, making it a valuable tool for research into immune checkpoints and potential cancer immunotherapies.

  • Reacts with mouse PD-1 (CD279).
  • Monoclonal antibody, clone J43.
  • Isotype: Armenian hamster IgG.
  • Purity: ≥95% (determined by SDS-PAGE).
  • Low endotoxin: ≤1EU/mg (≤0.001EU/μg).
  • Produced in an animal-free facility.
  • Sterile: 0.2 μm filtration.
  • Formulation: PBS, pH 6.5; contains no stabilizers or preservatives.

Catalog No. 50-562-100


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.