Learn More
Bio X Cell Antibody, Mouse PD-1, Clone RMP1-14, 5 MG
Supplier:  Bio X Cell BE0146R005MG
Encompass_Preferred
The InVivoMAb anti-mouse PD-1 (CD279) antibody is engineered for reliable in vivo blocking of PD-1 signaling. This research-grade antibody corresponds to the extensively published RMP1-14 clone, providing confidence for functional studies across various PD-1 applications.
  • RMP1-14 clone for in vivo blocking
  • InVivoMAb research-grade formulation
  • Available in a 5 mg size
Catalog No. 50-562-455