Learn More
Bio X Cell Antibody, anti-mouse PD-1, RMP1-14, IgG2a, 0.50 MG
Supplier: Bio X Cell BE0146R050MG
The RMP1-14 monoclonal antibody reacts with mouse PD-1 (CD279), a cell surface receptor crucial for immune regulation. It inhibits T-cell activation by blocking PD-L1 and PD-L2 binding, playing a key role in peripheral tolerance and cancer immunotherapy research.
- Reactivity: Mouse
- Clone: RMP1-14
- Isotype: Rat IgG2a, κ
- Applications: In vivo blocking of PD-1/PD-L signaling, in vitro organoids/organ-on-chip
- Purity: ≥95% (determined by SDS-PAGE)
- Sterility: 0.2 μm filtration
- Purification: Protein G
- Formulation: PBS, pH 7.0 (no stabilizers or preservatives)
- Endotoxin: ≤1EU/mg (≤0.001EU/μg)
- Molecular weight: 150 kDa
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.