Learn More
Bio X Cell PD-1 Antibody, RMP1-14, Rat IgG2a, 100 MG
Supplier: Bio X Cell BE0146R100MG
The RMP1-14 monoclonal antibody targets mouse PD-1 (CD279), a 50-55 kDa cell surface receptor. This antibody blocks the binding of both mouse PD-L1-Ig and PD-L2-Ig to PD-1, making it relevant for immunotherapies exploring anti-PD-1 mediated cancer treatments.
- Reacts with mouse PD-1 (CD279)
- Monoclonal antibody
- Blocks mouse PD-L1-Ig and PD-L2-Ig binding to PD-1
- Formulation: PBS, pH 7.0, no stabilizers or preservatives
- Endotoxin level: ≤1EU/mg (≤0.001EU/μg) via LAL assay
- Purity: ≥95% (SDS-PAGE)
- Sterility: 0.2 μm filtration
- Produced in animal-free facility, purified by Protein G
- Applications: in vivo blocking of PD-1/PD-L signaling, in vitro Organoids/Organ-on-Chip
- Storage: 4°C, do not freeze
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.