Learn More
Bio X Cell Antibody, Mouse PD-1 (CD279), RMP1-14, 25 MG
Supplier: Bio X Cell BE0146R025MG
The RMP1-14 monoclonal antibody targets mouse PD-1 (CD279), a 50-55 kDa cell surface receptor. It blocks binding of mouse PD-L1-Ig and PD-L2-Ig to PD-1, making it suitable for anti-PD-1 mediated immunotherapy studies.
- Target: mouse PD-1 (CD279), Clone: RMP1-14
- Reactivity: mouse
- Isotype: rat IgG2a, κ
- Formulation: PBS, pH 7.0 (no stabilizers/preservatives)
- Endotoxin: ≤1 EU/mg
- Purity: ≥95% (SDS-PAGE)
- Sterility: 0.2 μm filtration
- Molecular weight: 150 kDa
- Purification: protein G from animal-free facility
- Validated applications: in vivo PD-1/PD-L signaling blockade; in vitro organoids/organ-on-chip
- Storage: 4°C at stock concentration; do not freeze
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.