Learn More
Bio X Cell Monoclonal Antibody, mouse PD-L1 (B7-H1), 10F.9G2, Rat IgG2b, 5 MG
Supplier: Bio X Cell BE0101R005MG
This monoclonal antibody targets mouse PD-L1 (B7-H1/CD274), a 40 kDa type I transmembrane protein expressed on various immune and endothelial cells. The antibody blocks the interaction between PD-L1 and its receptors, PD-1 and B7-1 (CD80), thereby regulating T cell activation and potentially impacting tumor immune evasion.
- Targets mouse PD-L1 (B7-H1)
- Isotype: Rat IgG2b,
- Molecular weight: 150 kDa; Purity: 95%
- Sterile 0.2 m filtration
- Formulated in PBS, pH 6.5, without stabilizers or preservatives
- Purified from animal-free cell culture supernatant via protein G
- Reported applications: PD-L1 blockade (in vivo/in vitro), immunofluorescence, IHC (frozen), flow cytometry, western blot, in vitro organoids/organ-on-chip
- Store at stock concentration at 4C; Do not freeze
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.