Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bio X Cell Antibody, Clone 10F.9G2, Mouse PD-L1, 100 MG

Supplier:  Bio X Cell BE0101R100MG

Encompass_Preferred

The 10F.9G2 monoclonal antibody reacts with mouse PD-L1 (programmed death ligand 1), also known as B7-H1 or CD274. It blocks the interaction between PD-L1 and PD-1, and between PD-L1 and B7-1 (CD80), playing a crucial role in regulating immune responses and tumor immune evasion.

  • Clone: 10F.9G2
  • Target: Mouse PD-L1 (B7-H1, CD274)
  • Host species: Rat
  • Isotype: IgG2b, κ
  • Formulation: PBS, pH 6.5, no stabilizers or preservatives
  • Purity: ≥95% by SDS-PAGE
  • Sterility: 0.2 μm filtration
  • Production: Purified from cell culture supernatant in an animal-free facility
  • Purification: Protein G
  • Reported applications: In vivo PD-L1 blockade, in vitro PD-L1 blockade, immunofluorescence, immunohistochemistry (frozen), flow cytometry, western blot, in vitro organoids/organ-on-chip

Catalog No. 50-562-314


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.