Learn More
Bio X Cell Antibody, Thy1.2, 30H12, Rat IgG2b, 5 MG
Supplier: Bio X Cell BE0066R005MG
The 30H12 monoclonal antibody reacts with mouse Thy1.2 (CD90.2), a 25-35 kDa GPI-anchored membrane glycoprotein expressed across various cell types. It is particularly useful for depletion of T lymphocytes.
- Isotype: rat IgG2b, κ
- Reactivities: mouse
- Conjugation: unconjugated
- Immunogen: mouse thymus or spleen
- Formulation: PBS, pH 7.0, no stabilizers or preservatives
- Endotoxin: ≤1EU/mg (≤0.001EU/μg)
- Purity: ≥95% by SDS-PAGE
- Sterility: 0.2 μm filtration
- Production: purified from cell culture supernatant in animal-free facility
- Purification: protein G
- Molecular weight: 150 kDa
- Storage: store at stock concentration at 4°C, do not freeze
- Applications: in vivo ILC depletion, in vivo T cell depletion, Western blot
- Clone: 30H12
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.