Learn More
Bio X Cell Mouse TIM-3 Antibody, Clone RMT3-23, 5 MG
Supplier: Bio X Cell BE0115R005MG
The RMT3-23 monoclonal antibody reacts with mouse TIM-3 (CD366), a 60 kDa immune checkpoint receptor. It functions as a TIM-3 receptor antagonist, inhibiting signaling and suppressing tumor cell growth. This antibody is pivotal for research into immune response regulation and cancer immunotherapy.
- Clone: RMT3-23
- Reactivity: Mouse
- Isotype: Rat IgG2a, κ
- Immunogen: Recombinant mouse TIM-3
- Formulation: PBS, pH 7.0 (no stabilizers or preservatives)
- Endotoxin: ≤1 EU/mg (≤0.001 EU/μg)
- Purity: ≥95% by SDS-PAGE
- Sterility: 0.2 μm filtration
- Purification: Protein G
- Storage: Store at stock concentration at 4°C, do not freeze
- Molecular weight: 150 kDa
- Reported applications: in vivo TIM-3 neutralization, in vitro TIM-3 blocking, flow cytometry
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.