Learn More
Bio X Cell Monoclonal Antibody, anti-mouse VEGFR-2, DC101, 25 MG
Supplier: Bio X Cell BE0060R025MG
The DC101 monoclonal antibody targets mouse VEGFR-2 (vascular endothelial growth factor receptor 2), also known as CD309, KDR, and Flk-1. VEGFR-2 is a tyrosine protein kinase crucial for vascular development and permeability, highly expressed on endothelial cells. This antibody has been shown to inhibit VEGFR-2 signaling in vivo.
- Isotype: Rat IgG1, κ
- Immunogen: mouse VEGFR-2-SEAPs soluble receptor
- Formulation: PBS, pH 7.0, no stabilizers or preservatives
- Endotoxin: ≤1EU/mg (≤0.001EU/μg)
- Purity: ≥95% (SDS-PAGE)
- Sterility: 0.2 μm filtration
- Purification: protein G
- Reported applications: in vivo blocking of VEGF/VEGFR-2 signaling, in vitro blocking, western blot
- Storage: 4°C, do not freeze
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.