Learn More
Bio X Cell Antibody, anti-mouse VEGFR-2, Clone DC101, 5 MG
Supplier: Bio X Cell BE0060R005MG
The DC101 monoclonal antibody reacts with mouse VEGFR-2 (vascular endothelial growth factor receptor 2), also known as CD309, KDR, and Flk-1. VEGFR-2 is a tyrosine protein kinase family member playing key roles in vascular development and permeability upon binding to its ligand VEGF. The DC101 antibody inhibits VEGFR-2 signaling in vivo.
- Isotype: Rat IgG1, κ
- Conjugation: Unconjugated
- Immunogen: Mouse VEGFR-2-SEAPs soluble receptor
- Applications: In vivo VEGF/VEGFR-2 blocking, in vitro VEGFR blocking, Western blot
- Formulation: PBS, pH 7.0 (no stabilizers/preservatives)
- Endotoxin: ≤1EU/mg (≤0.001EU/μg)
- Purity: ≥95% (SDS-PAGE)
- Sterility: 0.2 μm filtration
- Production: Purified from animal-free cell culture supernatant
- Storage: 4°C; do not freeze
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.