Learn More
Sino Biological Anti-M.viridifaciens NA Antibody, Rabbit Polyclonal 20 µL
Catalog No. 5000750285
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Micromonospora viridifaciens Neuraminidase / Sialidase (Catalog#11679-V07E; BAA00852.1; Pro 42-Gly 403). Micromonospora viridifaciens Neuraminidase / Sialidase specific IgG was purified by Micromonospora viridifaciens Neuraminidase / Sialidase affinity chromatography.
Catalog No. 50-007-50285 Supplier Sino Biological Supplier No. 11679RP0220
Produced in rabbits immunized with purified, recombinant Micromonospora viridifaciens Neuraminidase / Sialidase (Catalog#11679-V07E; BAA00852.1; Pro 42-Gly 403). Micromonospora viridifaciens Neuraminidase / Sialidase specific IgG was purified by Micromonospora viridifaciens Neuraminidase / Sialidase affinity chromatography.