Learn More
enQuire Bioreagents Anti-MAP2 antibody | 25uL
Supplier:  enQuire Bioreagents QAB10310
Encompass_Preferred
Chicken polyclonal to MAP2. Reacts with: Mouse, Rat. Recombinant fragment corresponding to Human MAP2. Mix of recombinant human constructs of projection domain sequences, amino acids 235-1588.
Catalog No. NC1666503