Learn More
FUJIFILM Biosciences Anti MAP2 Guinea Pig 50uL
Catalog No. 5000450104
Encompass_Preferred
Microtubule-associated protein 2 MAP2 is a protein prevalent in neuronal cells It binds to and stabilizes microtubules1 There are four known isoforms of MAP2 the high molecular weight MAP2a and MAP2b around 280 kDa and the low molecular weight MAP2c and MAP2d around 70 kDa MAP2 is mainly expressed in the cell body and dendrites of neuronal cells and is widely used as a neuronal marker in the field of neuroscience The Anti MAP2 Guinea Pig is a guinea pig polyclonal antibody raised against MAP2 It can be used to perform multiplex immunohistochemistry Specific to MAP2
Catalog No. 50-004-50104 Supplier FUJIFILM Biosciences Supplier No. 01328551
Microtubule-associated protein 2 MAP2 is a protein prevalent in neuronal cells It binds to and stabilizes microtubules1 There are four known isoforms of MAP2 the high molecular weight MAP2a and MAP2b around 280 kDa and the low molecular weight MAP2c and MAP2d around 70 kDa MAP2 is mainly expressed in the cell body and dendrites of neuronal cells and is widely used as a neuronal marker in the field of neuroscience The Anti MAP2 Guinea Pig is a guinea pig polyclonal antibody raised against MAP2 It can be used to perform multiplex immunohistochemistry Specific to MAP2