Learn More
Charles River Labs Cell Solutions Anti-MART-1 (26-35) CD8+ T Cells, 1 to 2 Million cells/vial, Cryopreserved
Supplier:  Charles River Labs Cell Solutions ASTC1072
Encompass
MART-1 (Melanoma Antigen Recognized by T cells), also known as Melan-A, is a 118 amino acid long surface protein expressed mainly on melanocytes and most melanoma. This anti MART-1 T cells are CD8+ T cells with specificity to the naturally processed native MART-1 (27-35) 9-mer: AAGIGILTV. These T cells also recognize the analog peptide: ELAGIGILTV, a 10-mer (26-35) with L for A substitution in position 27
Catalog No. 50-225-6102
Only null left
Add to Cart
This item is not returnable. View return policy
Description
Description
Product Suggestions
Product Suggestions
Videos
Safety and Handling
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov