Learn More
MyBioSource Anti-MAT2A Antibody, Rabbit, 0.2ml
SDP
Supplier:  MyBioSource MBS60030830.2 ML
Encompass_Preferred
The Anti-MAT2A Antibody from MyBioSource is a rabbit polyclonal antibody produced by immunizing rabbits with a synthetic peptide corresponding to residues near the C-terminus of human MAT2A protein. This antibody is validated for WB, IHC, and ELISA applications. It is provided at 200 μg/ml in PBS with 0.02% sodium azide and 50% glycerol. The immunogen is a 17 amino acid peptide from near the carboxy terminus of human MAT2A. This antibody can detect MAT2A protein from human samples.
Catalog No. NC2610600