Learn More
Sino Biological Anti-MEK1 Antibody, Rabbit Polyclonal 100 µL
Catalog No. 5000750866
Encompass_Preferred
Produced in rabbits immunized with a synthetic peptide corresponding to the center region of the Human MEK1, and purified by antigen affinity chromatography.
Catalog No. 50-007-50866 Supplier Sino Biological Supplier No. 106830T36100
Produced in rabbits immunized with a synthetic peptide corresponding to the center region of the Human MEK1, and purified by antigen affinity chromatography.