Learn More
Sino Biological Anti-MEK2 Antibody, Rabbit Polyclonal 100 µL
Catalog No. 5000749737
Encompass_Preferred
Produced in rabbits immunized with a synthetic peptide corresponding to the center region of the Mouse MEK2, and purified by antigen affinity chromatography.
Catalog No. 50-007-49737 Supplier Sino Biological Supplier No. 107431T32100
Produced in rabbits immunized with a synthetic peptide corresponding to the center region of the Mouse MEK2, and purified by antigen affinity chromatography.