Learn More
Sino Biological Anti-MMAE Antibody (Biotin) 1 mg
Catalog No. 5000786247
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant F(ab')2 of human kappa light chain IgG, and then biotinylated.
Catalog No. 50-007-86247 Supplier Sino Biological Supplier No. 68218R0007B1
This antibody was obtained from a rabbit immunized with purified, recombinant F(ab')2 of human kappa light chain IgG, and then biotinylated.