Learn More
Sino Biological Anti-MMAE Antibody, Mouse MAb
Catalog No. 5000083716
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with MMAE conjugated to BSA. The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.
Catalog No. 50-000-83716 Supplier Sino Biological Supplier No. 68218MM0820
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with MMAE conjugated to BSA. The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.