Learn More
Sino Biological Anti-MMAE Antibody, Rabbit MAb 20 µg
Catalog No. 5000786243
Encompass_Preferred
This product is an antibody recombinantly expressed in HEK293 cells, and biotinylated via BirA enzyme.
Catalog No. 50-007-86243 Supplier Sino Biological Supplier No. 68218R000720
This product is an antibody recombinantly expressed in HEK293 cells, and biotinylated via BirA enzyme.