Learn More
BIOGEMS INTERNATIONAL INC Anti-Mouse CD115 (CSF-1R) FITC Size 25 g
SDP
Supplier:  BIOGEMS INTERNATIONAL INC 172125025
Encompass
The AFS98 monoclonal antibody specifically reacts with the mouse CD115 molecule, a 150 kDa receptor for the colony stimulating factor (CSF-1) or macrophage CFS (M-CFS), expressed by some epithelial cells, monocytes, osteoclasts, and macrophages. The colony stimulating factor-1 regulates the proliferation and the differentiation of the monocytic lineage cells.
The AFS98 antibody can be used to identify myeloid lineage cells.
Catalog No. 50-152-9830