Learn More
BIOGEMS INTERNATIONAL INC Anti-Mouse IL-13 SAFIRE Purified 50ug
SDP
Supplier:  BIOGEMS INTERNATIONAL INC 835122550
Encompass
The 1316H monoclonal antibody binds to mouse IL-13. IL-13 is an immunoregulatory cytokine produced primarily by activated Th2 cells, and also by mast cells and NK cells. Targeted deletion of IL-13 in mice resulted in impaired Th2 cell development and indicated an important role for IL-13 in the expulsion of gastrointestinal parasites. IL-13 exerts anti-inflammatory effects on monocytes and macrophages and it inhibits the expression of inflammatory cytokines such as IL-1β, TNF-α, IL-6 and IL-8. IL-13 has also been shown to enhance B cell proliferation and to induce isotype switching, resulting in increased production of IgE. Blocking of IL-13 activity inhibits the pathophysiology of asthma. The 1316H antibody is reported to be a neutralizing antibody.
Catalog No. 50-153-0441