Learn More
Seracare Life Sciences Inc ANTI-MONKEY IGA (A) AB
Supplier:  Seracare Life Sciences Inc 52100215
Encompass
Affinity purified polyclonal antibody to monkey IgA heavy chains (alpha) made in Goat. Product is in liquid form. Each lot is tested to assure specificity and lot-to-lot consistency using KPL's in-house ELISA assay.
Catalog No. 50-670-29