Learn More
Rockland Immunochemicals Anti-MONKEY IgM (mu chain) (GOAT) Antibody - 617-101-007
SDP
Supplier:  Rockland Immunochemicals 617101007
Encompass
Anti-Monkey IgM antibody specifically detects monkey IgM. Immunoglobulin M is the largest antibody isotype and the first to be secreted against an initial exposure to antigen. IgM is predominantly produced in the spleen. Formed from covalently linking 5 immunoglobulins together, the approximate molecular weight of IgM is 900kDa and possesses 10 binding sites (though due to the size of most antigens, not all sites are capable of binding at once). Due to this large size, IgM is typically isolated to the serum. Anti-Monkey IgM antibody is ideal for investigators in Immunology, Microbiology, and Cell Biology.
Catalog No. NC0156714