Learn More
Immunology Consultants anti-Monkeypox Virus A35R Antibody (Human - Clone #101) - Monoclonal
SDP
Catalog No. 5000435433
Encompass_Preferred
This human monoclonal antibody recognizes the A35R surface protein of Monkeypox virus, enabling precise detection in immunoassays. Its high specificity and strong binding affinity support research on viral structure, antigenicity, and diagnostic assay development. HMPX‑55A‑101 performs reliably in ELISA and Western blotting, offering clean signal and reproducible assay behavior.
Catalog No. 50-004-35433 Supplier Immunology Consultants Supplier No. HMPX55A101
This human monoclonal antibody recognizes the A35R surface protein of Monkeypox virus, enabling precise detection in immunoassays. Its high specificity and strong binding affinity support research on viral structure, antigenicity, and diagnostic assay development. HMPX‑55A‑101 performs reliably in ELISA and Western blotting, offering clean signal and reproducible assay behavior.