Learn More
Immunology Consultants anti-Monkeypox Virus B6R Antibody (Human - Clone #100) - Monoclonal
SDP
Catalog No. 5000435311
Encompass_Preferred
This human monoclonal antibody targets the B6R envelope protein of Monkeypox virus, providing high‑specificity detection for virology research and assay development. Its strong affinity supports sensitive detection in ELISA, Western blotting, and antigen‑capture formats. HMPX‑55A‑100 is well suited for studies of viral entry, immune response, and orthopoxvirus surveillance, offering consistent performance and low nonspecific binding across repeated assays
Catalog No. 50-004-35311 Supplier Immunology Consultants Supplier No. HMPX55A100
This human monoclonal antibody targets the B6R envelope protein of Monkeypox virus, providing high‑specificity detection for virology research and assay development. Its strong affinity supports sensitive detection in ELISA, Western blotting, and antigen‑capture formats. HMPX‑55A‑100 is well suited for studies of viral entry, immune response, and orthopoxvirus surveillance, offering consistent performance and low nonspecific binding across repeated assays