Learn More
Immunology Consultants anti-Monkeypox Virus M1R Antibody (Human - Clone #102) - Monoclonal
SDP
Catalog No. 5000435368
Encompass_Preferred
This monoclonal antibody targets the M1R membrane protein of Monkeypox virus, providing sensitive detection for virology research and antigen‑capture assays. Its strong specificity and low background support studies of viral assembly, immune recognition, and orthopoxvirus diagnostics. HMPX‑55A‑102 delivers consistent, high‑clarity performance across assay formats.
Catalog No. 50-004-35368 Supplier Immunology Consultants Supplier No. HMPX55A102
This monoclonal antibody targets the M1R membrane protein of Monkeypox virus, providing sensitive detection for virology research and antigen‑capture assays. Its strong specificity and low background support studies of viral assembly, immune recognition, and orthopoxvirus diagnostics. HMPX‑55A‑102 delivers consistent, high‑clarity performance across assay formats.