Learn More
ABclonal Technology MonoMethyl-Histone H3-K36 Polyclonal Antibody, 100UL, Validated in:WB, IHC, IF, IP, ChIP, ChIPseq
SDP
Supplier:  ABclonal Technology A2364
Encompass_Preferred
Immunogen: A synthetic monomethylated peptide around K36 of human histone H3 (NP_003520.1).•Host Species: Rabbit•Isotype: IgG•Species Reactivity:Human,Mouse,Rat,Other (Wide Range)•Validated Applications:WB,IF,IP,ChIP,ChIP-seq•Swiss Prot: Q16695•Gene ID:8290•Purification Method:Affinity purification•Storage: Store at -20℃. Avoid freeze / thaw cycles.|Buffer: PBS with 0.02% sodium azide,50% glycerol,pH7.3.This product is available in 20UL, 50UL, 100UL, and 200UL. Please contact customer service for pricing and availability on alternative sizes.
Catalog No. 50-130-7197