Learn More
BANGS LABORATORIES INC Quantum™ Simply Cellular anti-Mouse IgG, Versatile Flow Cytometry Application, Reliable Antibody Binding Capacity Assessment, 1 mL
Catalog No. NC1087345
Encompass
The Quantum™ Simply Cellular anti-Mouse IgG is a high-quality standard for quantifying cellular antigen expression and determining Antibody Binding Capacity (ABC) in flow cytometry. Its five populations of uniform microspheres enable accurate standard curves and instrument calibration, differentiating it through its Fc-specific capture and exceptional consistency for fluorescent applications. This product aids in evaluating detection thresholds and monitoring conjugated antibody stability.
Catalog No. NC1087345 Supplier BANGS LABORATORIES INC Supplier No. 815A
The Quantum™ Simply Cellular anti-Mouse IgG is a high-quality standard for quantifying cellular antigen expression and determining Antibody Binding Capacity (ABC) in flow cytometry. Its five populations of uniform microspheres enable accurate standard curves and instrument calibration, differentiating it through its Fc-specific capture and exceptional consistency for fluorescent applications. This product aids in evaluating detection thresholds and monitoring conjugated antibody stability.