Learn More
FUJIFILM Biosciences Anti Mouse Ago2 Monoclonal Ant
Catalog No. 5000450128
Encompass_Preferred
Ago2 Argonaute2 was isolated as one of the main components of RISC RNA-induced silencing complex AGO2 captures siRNA and microRNA which are working as a guide molecule for interaction with target mRNAs in RNAi pathway In this pathway AGO2 catalyzes the nicking of target mRNAs and binding between RISC and target mRNAs This product is a mouse monoclonal antibody 2D4 against Ago2 High specificity to mouse Ago2 Cross-reactivity with rat and hamster not cross-react with human Ago2
Catalog No. 50-004-50128 Supplier FUJIFILM Biosciences Supplier No. 01422023
Ago2 Argonaute2 was isolated as one of the main components of RISC RNA-induced silencing complex AGO2 captures siRNA and microRNA which are working as a guide molecule for interaction with target mRNAs in RNAi pathway In this pathway AGO2 catalyzes the nicking of target mRNAs and binding between RISC and target mRNAs This product is a mouse monoclonal antibody 2D4 against Ago2 High specificity to mouse Ago2 Cross-reactivity with rat and hamster not cross-react with human Ago2