Learn More
Bio X Cell InVivoMAb anti-mouse CD103, M290, 50 MG
Supplier:  Bio X Cell BE0026R050MG
Encompass_Preferred
The InVivoMAb anti-mouse CD103 (clone M290) is a functional antibody designed for research applications. This antibody specifically targets mouse CD103, an integrin protein vital for T cell interactions with epithelial cells and expressed on various immune cells including intraepithelial lymphocyte T cells, some regulatory T cells, and a subset of dendritic cells. It is validated for in vivo neutralization of CD103, as well as for immunofluorescence and flow cytometry.
  • Reacts specifically with mouse CD103.
  • Provided in a phosphate-buffered saline solution without preservatives.
  • Suitable for in vivo studies, capable of neutralizing CD103 activity.
  • High purity, with endotoxin levels suitable for sensitive applications.
  • Manufactured in an animal-free facility to minimize contaminants.
  • Stable for storage at refrigerated temperatures, ensuring product integrity.
Catalog No. NC2030216