Learn More
STEMCELL Technologies Anti-Mouse CD117 Antibody, Clone 2B8, PE, Size: 200 µg
SDP
Supplier:  STEMCELL Technologies 60025PE
Encompass
Rat monoclonal IgG2b antibody against mouse CD117 (c-Kit), PE-conjugated
Catalog No. 50-197-5123