Learn More
Fluidigm Corp ANTI-MOUSE CD11B-148ND
SDP
Supplier:  Fluidigm Corp 3148003B
Encompass
CD11b, also known as Mac-1, is a transmembrane glycoprotein known to be involved in cell adhesion and phagocytosis. CD11b is expressed on the surface of many leukocytes involved in the innate immune system, including monocytes, granulocytes, macrophages, dendritic cells, and natural killer cells.
Catalog No. 50-466-836