Learn More
STEMCELL Technologies Anti-Mouse CD11b Antibody, Clone M1/70, Alexa Fluor 488, Size: 100 µg
SDP
Supplier:  STEMCELL Technologies 60001AD
Encompass
Clone M1/70 is a rat monoclonal antibody that reacts with human, mouse, and rhesus CD11b, an ~170 kDa type 1 transmembrane glycoprotein which associates non-covalently with CD18 to form the heterodimeric Mac-1 receptor. Verified for use with flow cytometry assays, use this antibody in applications requiring the detection of CD11b. CD11b is expressed on the surface of granulocytes, monocytes, NK cells, dendritic cells, tissue macrophages, and subsets of T and B cells, and has been used as a marker to distinguish naïve and memory CD8+ T cells. The M1/70 antibody reportedly blocks iC3b binding to Mac-1. This antibody clone has been verified for purity assessments of cells isolated with EasySep™ kits, including EasySep™ Mouse Monocyte Isolation Kit (Catalog #19861). Visit STEMCELL.com for more information and related products.
Catalog No. 50-197-4863