Learn More
Fluidigm Corp ANTI-MOUSE CD11C-142ND
SDP
Supplier:  Fluidigm Corp 3142003B
Encompass
CD11c is a transmembrane glycoprotein also known as integrin αX or CR4. CD11c is primarily expressed on dendritic cells, NK cells, a subset of intestinal intraepithelial lymphocytes (IEL), and some activated T cells. It has also been associated with hairy cell leukemias, acute nonlymphocytic leukemias, and some B cell chronic lymphocytic leukemias.
Catalog No. 50-466-792