Learn More
STEMCELL Technologies Anti-Mouse CD138 (Syndecan-1) Antibody, Clone 281-2, PE, Size: 25 µg
SDP
Supplier:  STEMCELL Technologies 60035PE.1
Encompass
Clone 281-2 is a rat monoclonal antibody that reacts with an extracellular epitope on the core protein of mouse CD138 (Syndecan-1), a ~60 - 100 kDa type 1 transmembrane glycoprotein expressed on the surface of pre-B cells, immature B cells, and normal and malignant plasma cells (but not mature circulating B cells). Verified for use with flow cytometry and cell separation assays, use this antibody in applications requiring the detection of mouse CD138. CD138 is thought to act primarily as a receptor that modulates cell proliferation, cell migration and cell-matrix associations by linking the extracellular matrix to the cytoskeleton. This antibody clone has been verified for purity assessments of cells isolated with EasySep™ kits, including EasySep™ Mouse Pan-B Cell Isolation Kit (Catalog #19844) and EasySep™ Mouse CD19 Positive Selection Kit II (Catalog #18954). Visit STEMCELL.com for more information and related products.
Catalog No. 50-197-5220