Learn More
Biolegend PE/Dazzle™ 594 anti-mouse CD206 (MMR)
Supplier:  Biolegend 141731
Encompass_Preferred
PE/Dazzle™ 594 anti-mouse CD206 (MMR) [C068C2]; Isotype: Rat IgG2a, κ; Reactivity: Mouse; Apps: ICFC, FC; Size: 25 ug
Catalog No. NC0998574