Learn More
Fluidigm Corp ANTI-MOUSE CD25 (3C7)-151EU
SDP
Supplier:  Fluidigm Corp 3151007B
Encompass
CD25, a glycoprotein known as the low-affinity interleukin-2 receptor (IL-2R), enables T cells to respond to the growth-promoting cytokine IL-2. It is expressed on regulatory and activated T cells, activated B cells, macrophages, and monocytes.
Catalog No. NC0501729