Learn More
STEMCELL Technologies Anti-Mouse CD27 Antibody, Clone LG.3A10, Size: 500 µg
SDP
Supplier:  STEMCELL Technologies 60160
Encompass
The LG.3A10 antibody reacts with CD27, a 45 - 55 kDa member of the tumor necrosis factor receptor superfamily. CD27 is found on peripheral T cells, memory B cells, and NK cells and in mice has been found on most thymocytes, except CD46-CD8- precursors. CD27 and its ligand CD70 are involved in multiple aspects of T cell biology, including the costimulation and expansion of CD4+ and CD8+ T cells and differentiation to memory CD8+ T cells. CD27 interacts with TRAF family members, resulting in enhanced cellular proliferation and the induction of anti-apoptotic proteins. Additionally, CD27 plays important roles in T cell – B cell interactions and the maturation of effector T cells. The expansion of activated B cells and their differentiation to plasma cells is also reported to be enhanced by CD27.
Catalog No. 50-197-5751
Add to Cart
Description
Description
Product Suggestions
Product Suggestions
Videos
Safety and Handling
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov