Learn More
AbD Serotec ANTI MOUSE CD3:PE
Supplier:  AbD Serotec MCA500PE
Encompass
Rat Anti Mouse CD3: RPE, Monolconal, Clone: KT3, 100 Tests/1.0mL, Purified IgG conjugated to R. Phycoerythrin (RPE) - lyophilised, Isotype: IgG 2a (Rat), Store at +4 deg.C, Application: FC
Catalog No. 50-216-918