Learn More
Biolegend PE/Dazzle™ 594 anti-mouse CD335 (NKp46)
Supplier:  Biolegend 137630
Encompass_Preferred
PE/Dazzle™ 594 anti-mouse CD335 (NKp46) [29A1.4]; Isotype: Rat IgG2a, κ; Reactivity: Mouse; Apps: FC; Size: 100 ug
Catalog No. NC1116472