Learn More
STEMCELL Technologies Anti-Mouse CD3e Antibody, Clone 145-2C11, PE, Size: 200 µg
SDP
Supplier:  STEMCELL Technologies 60015PE
Encompass
Clone 145-2C11 is a hamster (Armenian) monoclonal antibody that reacts with the ~20 kDa CD3ε subunit of the mouse T cell receptor (TCR)/CD3 complex, which is expressed on the surface of circulating mature T cells and NKT cells, and variably on thymocytes. Verified for use with flow cytometry assays, use this antibody in applications requiring the detection of CD3e. The 145-2C11 antibody has been used for in vitro functional (blocking and activation) assays and has been reported to block binding by the clone 17A2 antibody. The 145-2C11 antibody is not recommended for use with formalin-fixed, paraffin-embedded sections. This antibody clone has been verified for purity assessments of cells isolated with EasySep™ kits, including EasySep™ Mouse T Cell Isolation Kit (Catalog #19851) and EasySep™ Mouse CD90.2 Positive Selection Kit II (Catalog #18951). Visit STEMCELL.com for more information and related products.
Catalog No. 50-197-5005