Learn More
STEMCELL Technologies Anti-Mouse CD48 (SLAMF2) Antibody, Clone HM48-1, APC, Size: 25 µg
SDP
Supplier:  STEMCELL Technologies 60162AZ.1
Encompass
The HM48-1 antibody reacts with mouse CD48, a 45 kDa GPI-anchored glycoprotein and a member of the signaling lymphocyte activation molecule (SLAM) family and Ig superfamily. SLAM family members play a role in multiple aspects of immune function including costimulation, cytotoxicity, and cytokine production. CD48 also plays a role in cell adhesion and T cell activation. In mice, CD48 is thought to be present on nearly all hematopoietic cells except neutrophils and ESLAM cells (CD45+EPCR+CD48-CD150+ hematopoietic stem cells with long-term self-renewal capacity). Ligands for CD48 include CD2 and CD244, binding of which helps to promote interactions between immune cells. HM48-1 is reported to be capable of blocking CD48:CD2 and CD48:CD244 interactions in vitro and in vivo. HM48-1 is also reported to inhibit the proliferative response of spleen cells to mitogens, inhibit CTL activity, and enhance the survival of cardiac allografts.
Catalog No. 50-197-5759
Add to Cart
Description
Description
Product Suggestions
Product Suggestions
Videos
Safety and Handling
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov