Learn More
Medchemexpress LLC Anti-Mouse CSF1R CD1 5mg | 5mg
SDP
Catalog No. 5000324622
Encompass_Preferred
Anti-Mouse (CSF1R/CD115) Antibody (AFS98) is an anti-mouse CSF1R IgG2a antibody inhibitor derived from rats Anti-Mouse (CSF1R/CD115) Antibody (AFS98) inhibits M-CSF-dependent colony formation and cell growth by blocking the binding of M-CSF to its receptor Anti-Mouse (CSF1R/CD115) Antibody (AFS98) can be used for the researches of inflammation metabolic disease cardiovascular disease and neurological disease such as such as stroke diabetes and arthritis[1][2][3][4]
Catalog No. 50-003-24622 Supplier Medchemexpress LLC Supplier No. HYP9901255MG
Anti-Mouse (CSF1R/CD115) Antibody (AFS98) is an anti-mouse CSF1R IgG2a antibody inhibitor derived from rats Anti-Mouse (CSF1R/CD115) Antibody (AFS98) inhibits M-CSF-dependent colony formation and cell growth by blocking the binding of M-CSF to its receptor Anti-Mouse (CSF1R/CD115) Antibody (AFS98) can be used for the researches of inflammation metabolic disease cardiovascular disease and neurological disease such as such as stroke diabetes and arthritis[1][2][3][4]