Learn More
Selleck Chemical LLC Anti-mouse CXCR3-InVivo
SDP
Catalog No. 503135866
Encompass_Preferred
Anti-mouse CXCR3-InVivo reacts with mouse CXCR3 also known as CD183 a 38 kDa chemokine receptor for CXCL9 (MIG) CXCL10 (IP-10) and CXCL11 (ITAC)
Catalog No. 50-313-5866 Supplier Selleck Chemical LLC Supplier No. A21475MG 20PC
Anti-mouse CXCR3-InVivo reacts with mouse CXCR3 also known as CD183 a 38 kDa chemokine receptor for CXCL9 (MIG) CXCL10 (IP-10) and CXCL11 (ITAC)