Learn More
Selleck Chemical LLC Anti-mouse CXCR3-InVivo
Supplier: Selleck Chemical LLC A21475MG 5PC
Anti-mouse CXCR3-InVivo reacts with mouse CXCR3 also known as CD183 a 38 kDa chemokine receptor for CXCL9 (MIG) CXCL10 (IP-10) and CXCL11 (ITAC)