Learn More
Selleck Chemical LLC Anti-mouse CXCR3-InVivo
SDP
Supplier:  Selleck Chemical LLC A21471MG
Encompass_Preferred
Anti-mouse CXCR3-InVivo reacts with mouse CXCR3 also known as CD183 a 38 kDa chemokine receptor for CXCL9 (MIG) CXCL10 (IP-10) and CXCL11 (ITAC)
Catalog No. 50-313-6168